Skip to Content
Merck
All Photos(1)

Key Documents

109525

Sigma-Aldrich

4-Methoxythiophenol

97%

Synonym(s):

4-Mercaptoanisole, 4-Methoxybenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4SH
CAS Number:
Molecular Weight:
140.20
Beilstein:
1446910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5831 (lit.)

bp

100-103 °C/13 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(S)cc1

InChI

1S/C7H8OS/c1-8-6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

NIFAOMSJMGEFTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxythiophenol was used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM′s effect on both n- and p-channel organic thin film transistors. 4-Methoxythiophenol was used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CsF-celite, an efficient solid state reagent for the syntheses of thioesters and thioethers.
Shah STA, et al.
Monatshefte fur Chemie / Chemical Monthly, 136(9), 1583-1589 (2005)
Modification of gold source and drain electrodes by self-assembled monolayer in staggered n-and p-channel organic thin film transistors.
Boudinet D, et al.
Organic Electronics, 11(2), 227-237 (2010)
Hadeel T Al-Sinjilawi et al.
Archiv der Pharmazie, 347(11), 861-872 (2014-09-16)
A series of substituted 4-oxopyrido[2,3-a]phenothiazine-3-carboxylic acids (6a-d) were prepared via cyclization of the corresponding ethyl 7-(arylthioxy)-8-nitro(or azido)-4-oxoquinoline-3-carboxylates (3a-d/4a-d), followed by hydrolysis of the resultant esters (5a-d). Among these tetracyclics, compound 6a with unsubstituted terminal benzo-ring D was the most active
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Articles

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service