Merck
  • Syntheses of metallo-pseudorotaxanes, rotaxane and post-synthetically functionalized rotaxane: a comprehensive spectroscopic study and dynamic properties.
Syntheses of metallo-pseudorotaxanes, rotaxane and post-synthetically functionalized rotaxane: a comprehensive spectroscopic study and dynamic properties.
Dalton transactions (Cambridge, England : 2003) (2017-08-05)
Saikat Santra, Somnath Bej, Mandira Nandi, Partha Mondal, Pradyut Ghosh
ABSTRACT
Herein, a bis-amido tris-amine macrocycle and five bipyridine-based bidentate chelating ligands were investigated towards various divalent transition metal ion (Ni
Sigma-Aldrich
1,7-Phenanthroline, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,5′-Dimethyl-2,2′-dipyridyl, 98%