Merck
  • The m6A demethylase FTO targets POLQ to promote ccRCC cell proliferation and genome stability maintenance.
The m6A demethylase FTO targets POLQ to promote ccRCC cell proliferation and genome stability maintenance.
Journal of cancer research and clinical oncology (2024-01-25)
Yichen He, Yimeng Chen, Zhengsheng Li, Changping Wu
ABSTRACT
As the first identified m6A demethylase, FTO has been implicated in the progression of various cancers. However, the specific mechanism of FTO in clear cell renal cell carcinoma (ccRCC) remains incompletely understood. In this study, we aimed to explore the potential molecular mechanisms influencing the progression of ccRCC. We initially assessed the expression of FTO in tumor and adjacent tissues using TCGA database, RT-qPCR, and Western blot. We then conducted CCK-8, cell cycle analysis, and colony formation assay to investigate the impact of FTO on ccRCC cell proliferation. MeRIP-seq and RNA-seq were employed to identify potential downstream targets of FTO in ccRCC, and these findings were further validated through dual-luciferase reporter assays and MeRIP-qPCR. Then, DNA damage and cell death were assessed separately through gammaH2AX immunofluorescence detection and the LIVE/DEAD Fixable Dead Cell Stain assay, respectively. Subsequently, we identified downstream pathways influenced by FTO's regulation of POLQ through TCGA database analysis and GSEA enrichment analysis. Validation was carried out through Western blot. FTO is highly expressed in ccRCC tissues and cell lines. Furthermore, ROC curve demonstrates that FTO contributes to the diagnosis of ccRCC. FTO modulates m6A modification, consequently influencing the expression of POLQ, thus facilitating cell proliferation and maintaining genome stability in ccRCC. FTO could potentially serve as a diagnostic marker for ccRCC. FTO promotes the progression of ccRCC by regulating m6A modification, making the inhibition of FTO a potential novel therapeutic strategy in ccRCC.
Sigma-Aldrich
Monoclonal Anti-POLQ antibody produced in mouse, clone 1C11, purified immunoglobulin, buffered aqueous solution