Merck
  • Comparative effects of incretin-based therapy on early-onset diabetic nephropathy in rats: Role of TNF-α, TGF-β and c-caspase-3.
Comparative effects of incretin-based therapy on early-onset diabetic nephropathy in rats: Role of TNF-α, TGF-β and c-caspase-3.
Life sciences (2021-05-19)
Heba A Habib, Gehan H Heeba, Mohamed M A Khalifa
ABSTRACT
Diabetic nephropathy, a major threat to diabetic patients, is considered as the main reason for end-stage renal disease. Fortunately, incretin-based therapy has been aroused as considerable source to attenuate diabetic renal damage. This study aimed to investigate whether superior protective effects on the progression of diabetic kidney are exerted by glucagon-like peptide-1 analog, exenatide, or dipeptidyl peptidase-4 inhibitor, sitagliptin. Male Wistar rats were fed high-fat diet for 2 weeks followed by injection of low dose streptozotocin to induce type 2 diabetes mellitus. Four weeks after induction of diabetes, diabetic rats were administered vehicle, exenatide (5 μg/kg/day, SC) or sitagliptin (10 mg/kg/day, orally) for 4 weeks. Different incretin mimetic agents improved renal function as evident by significant decreases in serum creatinine and urea levels with decline in urinary microalbuminuria and marked improvement in histological alterations. Both treated diabetic rats also exhibited a significant improvement in metabolic intolerance with more pronounced effect of exenatide on glucose regulation. Ameliorated renal oxidative stress alongside significant downregulation in transforming growth factor-beta, tumor necrosis factor-alpha and cleaved-caspase-3 protein expressions in renal tissues were recorded in treated diabetic rats. Administration of either exenatide or sitagliptin showed ameliorative effects on early diabetic nephropathy without notable differences between their renal protective effects. However, further clinical studies are still required to ensure their comparative promising effects on the management of renal complication of diabetes.
Sigma-Aldrich
Sitagliptin, ≥98% (HPLC)