Skip to Content
Merck
All Photos(1)

Key Documents

51343-U

MXT Valves for Fast Chromatography

stainless steel, pkg of 1 ea

Synonym(s):

Fast Chromatography Valves

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51
Pricing and availability is not currently available.

material

stainless steel

description

MXT715-000 2-position, 6-port

packaging

pkg of 1 ea

General description

The MXT715-000 2-position, 6-port switching valve is ideal for use as a two-column switching valve, enabling the same system to be used with more than one column to easily accommodate multiple users and applications. It can also be used in a traditional injection valve configuration and offers the Rheodyne patented MMB (Make-Before Break) feature for improved reproducibility and system stability.

Application


  • Investigation of high-speed gas chromatography using synchronized dual-valve injection and resistively heated temperature programming: This research utilizes MXT Valve technology in a high-speed gas chromatography setup, demonstrating significant advancements in analytical chemistry. The study focuses on the synchronized dual-valve injection system that enhances the efficiency and speed of chromatographic separations, making it a vital tool for complex mixture analyses in both academic and industrial settings. This application aligns with the need for rapid and precise chemical analysis, essential in fields ranging from pharmaceuticals to environmental science (Reid et al., 2007).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service