Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200745

Sigma-Aldrich

Anti-Protein A antibody, Mouse Monoclonal

clone SPA-27, purified from hybridoma cell culture

Synonym(s):

Anti-IgG binding protein A, Anti-SpA, Anti-Staphylococcal protein A, Anti-Staphylococcus ayreus protein A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
SPA-27, monoclonal
application:
DB
ELISA (i)
FACS
IF
IP
species reactivity:
Staphylococcus aureus
technique(s):
dot blot: suitable
flow cytometry: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunoprecipitation (IP): 2.5-5 μg/test using purified protein A from Staphylococcus aureus
indirect ELISA: 0.25-0.5 μg/mL using 10 μg/mL of protein A from Staphylococcus aureus for coating. To reduce nonspecific binding we recommend on detection using a secondary anti-mouse, F(ab’)2 fragment antibody
citations:
2

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SPA-27, monoclonal

form

buffered aqueous solution

mol wt

~42 kDa

species reactivity

Staphylococcus aureus

concentration

~1.0 mg/mL

technique(s)

dot blot: suitable
flow cytometry: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunoprecipitation (IP): 2.5-5 μg/test using purified protein A from Staphylococcus aureus
indirect ELISA: 0.25-0.5 μg/mL using 10 μg/mL of protein A from Staphylococcus aureus for coating. To reduce nonspecific binding we recommend on detection using a secondary anti-mouse, F(ab’)2 fragment antibody

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Staphylococcus aureus protein A, also known as SpA or IgG binding protein A, is a 42 kDa single chain polypeptide isolated from the cell wall of S. aureus Cowan I strain. It consists of a single polypeptide chain containing five repetitive immunoglobulin-binding domains, rich in aspartic and glutamic acids.Anti-Protein A antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the SPA-27 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c immunized mouse.

Specificity

Monoclonal Anti-Protein A specifically recognizes Protein A from Staphylococcus aureus bacteria.

Immunogen

Protein A from Staphylococcus aureus Cowan I strain

Application

The antibody may be used in various immunochemical techniques including immunoblotting, dot blot, ELISA, immunofluorescence, immunoprecipitation and flow cytometry (FACS).

Biochem/physiol Actions

Protein A is considered as a universal reagent in biochemistry and immunology, due to its affinity for the Fc region of many mammalian immunoglobulins. It is used for different applications such as purification of immunoglobulins by affinity chromatography, cell surface studies, radioimmunoassay (RIA), enzyme immunoassay (EIA), immunoprecipitations and many other procedures. It can be used either in its native form, conjugated to various markers or serve as a tag for a specific protein of interest. In addition, protein A has low affinity to mouse monoclonal antibodies from IgG1 isotype. Thus, a monoclonal anti-protein A antibody of the mouse IgG1 isotype has an advantage due to its specific targeting and binding to protein A via its Fab fragment. Monoclonal anti-protein A antibody can serve a useful tool for quantification and visualization of S. aureus bacteria and bacterial contaminations in different examined samples and for detection of protein A tagged proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

This product is for R&D use only, not for drug, household, or other uses. Please consult the material safety data sheet (MSDS) for information regarding hazards and safe handling practices.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spt6 is required for the fidelity of promoter selection
Doris S, et al.
Molecular Cell, 72(4), 687-699 (2018)
M Graille et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(10), 5399-5404 (2000-05-11)
Staphylococcus aureus produces a virulence factor, protein A (SpA), that contains five homologous Ig-binding domains. The interactions of SpA with the Fab region of membrane-anchored Igs can stimulate a large fraction of B cells, contributing to lymphocyte clonal selection. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service