CLS3912
Corning® 96 Well Solid Polystyrene Microplate
flat bottom, white polystyrene, bag of 25, non-sterile, lid: no
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
About This Item
white bottom
white polystyrene
Recommended Products
material
flat bottom
white bottom
white polystyrene
sterility
non-sterile
feature
lid: no
skirt
packaging
bag of 25
manufacturer/tradename
Corning 3912
size
96 wells
well volume
360 μL
working volume
75-200 μL
color
white
suitability
suitable for (cell cuture)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
- Without lids, lids supplied separately
- White microplates enhance luminescent signals and have low background luminescence and fluorescence
- Designed to reduce well-to-well crosstalk
- Total volume is 360μL; recommended working volume of 75 to 200μL
- Conforms to standard microplate footprint and dimensions
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service