Skip to Content
Merck
All Photos(1)

Key Documents

PHR1673

Supelco

Lidocaine Related Compound H

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N-(2,6-Dimethylphenyl)chloroacetamide, n-(2,6-Dimethylphenyl)chloroacetamide, 1-Chloroacetylamino-2,6-dimethylbenzene, 2-Chloro-2′,6′-acetoxylidide, 2-Chloro-2′,6′-dimethylacetanilide, 2-Chloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2-chloroacetamide, N-(2,6-Xylyl)chloroacetamide, N-Chloroacetyl-2,6-dimethylaniline, Chloroacetamido-2,6-xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONHC6H3(CH3)2
CAS Number:
Molecular Weight:
197.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1366080

API family

lidocaine

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-151 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cc1cccc(C)c1NC(=O)CCl

InChI

1S/C10H12ClNO/c1-7-4-3-5-8(2)10(7)12-9(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)

InChI key

FPQQSNUTBWFFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Lidocaine Related Compound H is one of the impurities of the local anesthetic lidocaine found in bulk powder and dosage forms.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Simultaneous estimation of lidocaine hydrochloride, tribenoside, and their related compounds in pharmaceutical formulations by high-performance liquid chromatography (HPLC) combined with a UV/Vis detector
  • Multi-analysis of dexpanthenol, lidocaine hydrochloride, mepyramine maleate, and their related impurities in their combined dosage forms by high-performance liquid chromatography (HPLC)-diode array detection
  • Development of an HPLC method for the estimation of three active pharmaceutical ingredients along with their impurities in a semi-solid topical formulation

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2078 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina Plenis et al.
Chromatographia, 76(5-6), 255-265 (2013-03-14)
A new liquid chromatographic (LC) method for simultaneous determination of lidocaine hydrochloride (LH) and tribenoside (TR) along with their related compounds in pharmaceutical preparations is described. Satisfactory LC separation of all analytes after the liquid-liquid extraction (LLE) procedure with ethanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service