Skip to Content
Merck
All Photos(1)

Key Documents

NIST870

Column performance test mixture of liquid chromatography

NIST® SRM® 870

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.1 mL

manufacturer/tradename

NIST®

technique(s)

liquid chromatography (LC): suitable

application(s)

general analytical

format

multi-component solution

General description

SRM 870 is composed of a blend of five organic compounds in methanol: uracil, toluene, ethyl benzene, quinizarin, and amitriptyline added as amitriptyline hydrochloride. One unit of SRM 870 contains five ampoules, each containing about 1.1 mL of the mixture. For more information, please refer to the SDS and COA.
SRM 870_cert

SRM 870_SDS

Application

The Standard Reference Material is intended tocharacterize various aspects of liquid chromatographic (LC) column performance,such as efficiency, void volume, methylene selectivity, retentiveness, andactivity toward chelators and organic bases. It can also be used for columnclassification to assist in column selection during method development, as acontrol material for monitoring LC column performance over time, and in qualitycontrol for column manufacturing.
The Standard Reference Material is intended to characterize various aspects of liquid chromatographic (LC) column performance, such as efficiency, void volume, methylene selectivity, retentiveness, and activity toward chelators and organic bases. It can also be used for column classification to assist in column selection during method development, as a control material for monitoring LC column performance over time, and in quality control for column manufacturing.

Features and Benefits

  • Information values for detection response forvarious components are provided.
  • Expiration details, instructions for use,handling, and storage are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Uracil (C4H4N2O2), Toluene (C6H5CH3), Ethyl Benzene (C8H10), Quinizarin (C14H8O4), Amitriptyline HCl (C20H24ClN), Methanol (CH3OH)

Reference/Informational value is provided for the following analytes.
Uracil (C4H4N2O2), Toluene (C6H5CH3), Ethyl Benzene (C8H10), Quinizarin (C14H8O4), Amitriptyline HCl (C20H24ClN), Methanol (CH3OH)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service