Skip to Content
Merck
All Photos(1)

Key Documents

NIST3255

Camellia sinensis (green tea) extract

NIST® SRM® 3255

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1 g

manufacturer/tradename

NIST®

format

matrix material

General description

One unit of SRM 3255 comprises five packets, each containing approximately 1 g of extract. For more information, plese refer to the SDS and COA.

SRM 3255_cert

SRM 3255_SDS

Application

The Standard Reference Material (SRM) is primarily intended for the validation of analytical methods used to determine catechins, xanthines, and elements in extracts of green tea (Camellia sinensis) and similar matrices. It can also be utilized for quality assurance, particularly when assigning values to in-house control materials.

Features and Benefits

  • Certified mass fraction values for catechins and xanthines are reported on a dry-mass basis and values are metrologically traceable to the SI unit of mass fraction (expressed as milligrams per gram).
  • Reference mass fraction values for epigallocatechin methylgallate, gallic acid, theanine, and theophylline and elements in green tea are provided.
  • Maintenance and expiration of SRM certificate, instructions for use and storage are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: (–)-Epicatechin, (–)-Epicatechin gallate, (–)-Epigallocatechin, (–)-Epigallocatechin gallate, (–)-Epigallocatechin methylgallate, (–)-Gallocatechin, (–)-Gallocatechin gallate, (+)-Catechin, Caffeine, Gallic acid, L-Theanine, Theobromine, Theophylline
Trace Elements: Aluminum (Al), Arsenic (As), Copper (Cu), Iron (Fe), Lead (Pb), Manganese (Mn), Zinc (Zn)

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service