Skip to Content
Merck
All Photos(1)

Key Documents

03627

Supelco

Diquat dibromide-(ethylene-d4)

PESTANAL®, analytical standard

Synonym(s):

1,1′-Ethylene-d4-2,2′-dipyridylium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D4H8Br2N2
Molecular Weight:
348.07
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥98 atom % D

product line

PESTANAL®

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

[2H]C1([2H])C([2H])([2H])[N+]2=CC=CC=C2C3=[N+]1C=CC=C3.C

Related Categories

General description

Diquat dibromide-(ethylene-d4) is a post-emergence and nonselective pesticide widely used for weed control management in agricultural crops and fruit orchards.

Application

Diquat dibromide-(ethylene-d4) may be used as a standard for quaternary ammonium pesticide analyses in food and beverage samples using cation-exchange liquid chromatography coupled with high-resolution mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 3

Target Organs

Eyes, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast analysis of quaternary ammonium pesticides in food and beverages using cation?exchange chromatography coupled with isotope?dilution high?resolution mass spectrometry.
Nardin T, et al.
Journal of Separation Science (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service