Skip to Content
Merck
All Photos(1)

Key Documents

MABC1593

Sigma-Aldrich

Anti-CRBN Antibody, clone 4D6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
4D6 (-3H10-D6)
monoclonal
application:
IP
WB
species reactivity:
human, mouse, human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:

antibody form

purified antibody

clone

4D6 (-3H10-D6)
monoclonal

purified by

using protein G

species reactivity

human, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

General description

His-tagged recombinant protein fragment corresponding to the N-terminal region of human Cereblon.

Specificity

Clone 4D6 (-3H10-D6) is a mouse monoclonal antibody that detects human Cereblon. It targets an epitope within the N-terminal domain.

Application

Western Blotting Analysis: 20 ug from a representative lot detected CRBN in MOLT4 wild type cell lysates (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CRBN in MOLT4 wild type cell lysates (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna Austria).

Quality

Evaluated by Western Blotting in MOLT-4 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Cereblon.(CRBN) in MOLT-4 cell lysate.

Physical properties

~50 kDa observed; 50.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service