Skip to Content
Merck
All Photos(1)

Key Documents

HX0600

Supelco

Hydrobromic Acid

48%, Meets ACS Specifications GR ACS

Synonym(s):

Hydrogen Bromide Solution, Hydrobromic Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
concentration:
47.0-49.0% HBr

Product Name

Hydrobromic Acid 48%, Meets ACS Specifications GR ACS

vapor pressure

10.6 hPa ( 20 °C)

Quality Level

form

liquid

concentration

47.0-49.0% HBr

impurities

≤0.001% Phosphate (PO4)
≤0.003% Iodide (I)
≤0.003% Sulfate and sulfite (as SO4)
≤0.01 ppm Selenium (Se)
≤0.05% Chloride (Cl)
≤1 ppm Iron (Fe)
≤5 ppm Heavy metals (as Pb)

ign. residue

≤0.002%

color

yellow

bp

126 °C/1013 hPa

mp

-11 °C

shipped in

ambient

storage temp.

room temp

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay: 47.0-49.0%
Chloride (Cl): 0.05% max
Color: Yellow
Form: Liquid
Heavy metals (as Pb): 5ppm max
Iodide (I): 0.003% max
Iron (Fe): 1ppm max
Phosphate (PO4): 0.001% max
Residue after ignition: 0.002% max
Selenium (Se): 0.01ppm max
Sulfate and sulfite (as SO4): 0.003% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service