Skip to Content
Merck
All Photos(1)

Key Documents

8.05797

Sigma-Aldrich

Malondialdehyde bis (diethyl acetal)

for synthesis

Synonym(s):

Malondialdehyde bis (diethyl acetal), 1,1,3,3-Tetraethoxypropane, Malondialdehyde tetraethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHCH2CH(OC2H5)2
CAS Number:
Molecular Weight:
220.31
MDL number:
UNSPSC Code:
12352112
EC Index Number:
204-533-1
form:
liquid
Assay:
≥95.0% (GC)

Quality Level

Assay

≥95.0% (GC)

form

liquid

potency

1610 mg/kg LD50, oral (Rat)

bp

220 °C/1013 hPa

mp

-90 °C

transition temp

flash point 78 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H24O4/c1-5-12-10(13-6-2)9-11(14-7-3)15-8-4/h10-11H,5-9H2,1-4H3

InChI key

KVJHGPAAOUGYJX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.916 - 0.918
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service