Skip to Content
Merck
All Photos(1)

Key Documents

528824

Sigma-Aldrich

PARP Inhibitor XIV

The PARP Inhibitor XIV, also referenced under CAS 1104546-89-5, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor XIV, 4-(1-Methyl-1H-pyrrol-2-ylmethylene)-4H-isoquinoline-1,3-dione, BYK204165

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]1(c(ccc1)\C=C2\c3c(cccc3)C(=O)NC\2=O)C

InChI

1S/C15H12N2O2/c1-17-8-4-5-10(17)9-13-11-6-2-3-7-12(11)14(18)16-15(13)19/h2-9H,1H3,(H,16,18,19)/b13-9-

InChI key

BTYSIDSTHDDAJW-LCYFTJDESA-N

General description

A cell-permeable isoquinolinedione compound that potently inhibits PARP-1 activity (pIC50 = 7.35; IC50 = 45 nM) in a NAD+-competitive and reversible manner, while affecting PPAR-2 activity with a 100-fold lower potency. Selectively inhibits PARP-1-mediated cellular PAR formation and is of little effect against PAR formation in fibroblasts derived from parp-1(-/-) mice. Its in vivo application is limited by a poor aqueous solubility (~9 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Eltze, T., et al. 2008. Mol. Pharmacol.74, 1587.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service