Skip to Content
Merck
All Photos(1)

Key Documents

251500-M

Sigma-Aldrich

DAF-2

A highly sensitive and specific fluorescent indicator for the direct detection of nitric oxide (NO) in vitro.

Synonym(s):

DAF-2, 4,5-Diaminofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14N2O5
CAS Number:
Molecular Weight:
362.34
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

fluorescence

λex ~495 nm
λem ~515 nm

storage temp.

−20°C

SMILES string

Nc1cc2c(cc1N)C(=O)OC32c4c(cc(cc4)O)Oc5c3ccc(c5)O

InChI

1S/C20H14N2O5/c21-15-7-11-14(8-16(15)22)20(27-19(11)25)12-3-1-9(23)5-17(12)26-18-6-10(24)2-4-13(18)20/h1-8,23-24H,21-22H2

InChI key

LTYUPYUWXRTNFQ-UHFFFAOYSA-N

General description

A highly sensitive and specific fluorescent indicator for the direct detection of nitric oxide (NO) in vivo. The relatively non-fluorescent DAF-2 reacts rapidly with NO in the presence of oxygen to yield the highly fluorescent compound triazolofluorescein (DAF-2 T). Can detect NO under neutral conditions (detection limit = 5 nM). Excitation maximum: 495 nm; emission maximum: 515 nm.

Biochem/physiol Actions

Primary Target
Fluorescent indicator for the direct detection of nitric oxide (NO) in vitro.

Warning

Toxicity: Irritant (B)

Physical form

A 5 mM (1 mg in 550 µl) solution in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Berkels, R., et al. 2000. Cell Calcium27, 281.
Kojima, H., et al. 1998. Anal. Chem. 70, 2446.
Kojima, H., et al. 1998. Chem. Pharm. Bull. 46, 373.
Nakatsubo, N., et al. 1998. FEBS Lett. 427, 263.

Legal Information

Manufactured by Daiichi Pure Chemicals Co., Ltd
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service