Skip to Content
Merck
All Photos(1)

Key Documents

1.14731

Supelco

Hydrogen Peroxide Test, photometric

photometric, 2.0-20.0 mg/L (H2O2), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Hydrogen Peroxide Cell Test, photometric, 2.0-20.0 mg/L (H2O2), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

hydrogen peroxide

measuring range

0.25-5.0 mg/L (H2O2)
2.0-20.0 mg/L (H2O2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Titanyl sulfate)

storage temp.

15-25°C

General description

Method: photometric 2.0 - 20.0 mg/l H₂O₂ Spectroquant®

The method is analogous to DIN 38409-15.

This Spectroquant® Hydrogen Peroxide Cell Test allows the accurate quantification of the hydrogen peroxide concentration in disinfectant and rinsing solutions.

Method applied: In a solution acidified with sulfuric acid, hydrogen peroxide and compounds containing hydrogen peroxide react with a titanic acid ester to form yellow peroxotitanic acids which are determined photometrically.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Tests are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Protocols

Preparation of a standard solution for Hydrogen peroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service