Skip to Content
Merck
All Photos(1)

Key Documents

1.09214

Millipore

Adapter for contact plates

For use with MAS-100 NT® / MAS-100 VF®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74
Pricing and availability is not currently available.

material

aluminum assembly

Agency

non-compendial

description

For use with MAS-100 NT and MAS-100 VF
connector

product line

MAS-100®

feature

autoclavable

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

storage temp.

no temp limit

General description

The MAS-100®portable air samplers can be used with commercially available contact plates. For MAS-100 NT® and MAS-100 Eco® instruments, the contact plate adapter allows the central positioning of contact plates on the plate holder to guarantee that all impact holes will be on the agar surface. For MAS-100 VF® instruments, no adapter is required for a 180-degree rotation and positioning of the three clamps is sufficient to fix the culture plate in the middle. The contact plate adapter comes with three blue hexagon socket screws and an Allen wrench for attachment to the plate holder. Due to the narrower diameter and greater height of the contact plates, a specially designed perforated lid, such as 1191490001, which fits to lockable and non-lockable ICR/ICRplus contact plates, is also required.

Application

Adapter for contact plates is an additional accessory for Air Sampler that is used for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms using contact plates instead of 90 mm settleplates. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG
MAS-100 VF is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service