Key Documents
CLS999815
Corning® phenolic cap, PTFE liner
15-415 thread
Synonym(s):
Corning cellstack cell culture chambers, black phenolic, phenolic caps, screw caps, tissue culture chambers, Corning® 9998
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
Pricing and availability is not currently available.
material
PTFE liner
black phenolic cap
manufacturer/tradename
Corning 9998-15
H
~16 mm
fitting
15-415 thread
General description
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)