Skip to Content
Merck
All Photos(1)

Key Documents

805777

Sigma-Aldrich

2-Ketobutyric-1-13C acid

≥99 atom % 13C, ≥97% (CP)

Synonym(s):

α-Ketobutyric-1-13C acid, 2-Oxobutanoic-1-13C acid, 2-Oxobutyric-1-13C acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC3H6O3
Molecular Weight:
103.08
UNSPSC Code:
12352106
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99 atom % 13C

Assay

≥97% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+1

storage temp.

−20°C

SMILES string

CCC([13C](O)=O)=O

InChI

1S/C4H6O3/c1-2-3(5)4(6)7/h2H2,1H3,(H,6,7)/i4+1

InChI key

TYEYBOSBBBHJIV-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service