Merck
Pentacene thin-film transistors encapsulated by a thin alkane layer operated in an aqueous ionic environment.
Advanced materials (Deerfield Beach, Fla.) (2010-08-25)
Martin Göllner, Martin Huth, Bert Nickel
RESUMEN
The encapsulation of pentacene thin-film transistors (TFTs) by a 50 nm thick layer of a long chain alkane (C₄₄H₉₀) enables the operation in an aqueous solution of 1 mM NaCl. It is the first time that pentacene TFTs operate successfully in a liquid environment. This opens new perspectives for biocompatible sensor devices based on pentacene TFTs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tetratetracontane, 99%
Supelco
Tetratetracontane, analytical standard