Documentos clave
Z740417
Specimen containers with snap cap
size 300 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polypropylene cap
polypropylene container
packaging
pack of 100 ea
manufacturer/tradename
Simport C580-3
size
300 mL
General description
These disposable containers made of polypropylene, are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting