Documentos clave
Z544353
Molymod® bonding links
Flex bond, purple
About This Item
Código UNSPSC:
60104506
NACRES:
NB.17
envase
pkg of 25
pkg of 4 pkg
fabricante / nombre comercial
Molymod
Longitud
35 mm
color
purple
Descripción general
The bond lengths stated in the table are measured from atom to atom. Bonding links are available in various colors and may be used to represent different bond types. For example, gray links can represent covalent bonds, while purple or white links can represent ionic bonds.
Información legal
Molymod is a registered trademark of Spiring Enterprises Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)