Merck
  • In situ formation of β-glycosyl imidinium triflate from participating thioglycosyl donors: elaboration to disarmed-armed iterative glycosylation.
In situ formation of β-glycosyl imidinium triflate from participating thioglycosyl donors: elaboration to disarmed-armed iterative glycosylation.
Chemical communications (Cambridge, England) (2012-10-02)
Yu Hsien Lin, Bhaswati Ghosh, Kwok-Kong Tony Mong
ABSTRACT
β-Glycosyl imidinium triflate is generated from participating thioglycoside donors for disarmed-armed iterative glycosylations and one-pot oligosaccharide synthesis.
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)