This product is manufactured by PhytoLab GmbH & Co. The material is of completely synthetic origin.
PHL83261
trans-Pterostilbene
phyproof® Reference Substance
Synonym(s):
Pterostilbene, 3,5-Dimethoxy-4′-hydroxystilbene, 4-[(1E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥98.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
COc1cc(OC)cc(\C=C\c2ccc(O)cc2)c1
InChI
1S/C16H16O3/c1-18-15-9-13(10-16(11-15)19-2)4-3-12-5-7-14(17)8-6-12/h3-11,17H,1-2H3/b4-3+
InChI key
VLEUZFDZJKSGMX-ONEGZZNKSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Biochem/physiol Actions
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can you please provide me with the origin of the trans-pterosilbene, and is it derived from natural product or synthesized?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service