27142
Screw cap, solid top with aluminum liner, pkg 100
black phenolic solid cap, aluminum liner, for use with 4 mL vial with 13-425 thread
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
aluminum liner
black phenolic solid cap
feature
closure type black phenolic solid top cap
packaging
pkg of 100 ea
parameter
135 °C temp. limit
fitting
thread for 13-425
compatibility
for use with 4 mL vial with 13-425 thread
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Screw caps are available as solid caps with aluminum liner. Caps are made from phenolic resin. Solid phenolic caps can withstand a maximum temperature of 205° C.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service