26270-U
SGE Needles for luer lock syringes
needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm, point style, 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
23 ga
description
NLL-5/23
feature
point style 2
packaging
pkg of 5 ea
manufacturer/tradename
SGE 39802
needle L × O.D.
50 mm × 0.63 mm
fitting
luer lock
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SGE Needles for luer lock syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service