26239
SGE Plunger-Protected Syringe, Fixed Needle
volume 10 μL, needle size 26 ga, needle L 50 mm
About This Item
Recommended Products
material
bevel tip stainless steel needle
needle size
26 ga
description
10F
feature
point style #2
packaging
pkg of 10 ea
manufacturer/tradename
SGE 002033
needle L
50 mm
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | SML3664 | SML3329 | SML3463 |
---|---|---|---|
≥98% (HPLC) | ≥98% (HPLC) | ≥98% (HPLC) | ≥98% (HPLC) |
100 | 100 | 100 | 100 |
-10 to -25°C | -10 to -25°C | -10 to -25°C | -10 to -25°C |
DMSO: 2 mg/mL, clear | DMSO: 2 mg/mL, clear | DMSO: 2 mg/mL, clear | DMSO: 2 mg/mL, clear (Warmed) |
, White to very dark grey | white to beige | white to beige | , Red to very dark purple |
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service