22478
Supeltex M-4 Packed Column Ferrule, 1/4 in. Column O.D.
I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 50 ea
All Photos(1)
About This Item
UNSPSC Code:
41121815
Recommended Products
packaging
pkg of 50 ea
I.D.
1/4 in.
fitting
1/4 in. GC fitting ferrule for 1/4 in. O.D. packed column
compatibility
1/4 in. O.D. Column
General description
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)