packaging
pkg of 1 ea
Related Categories
General description
Correct fitting of a ferrule is very important in column performance. The ferrule needs to be properly sized and squarely cut using a tubing cutter and the cut should be examined by a magnifier. This is important as column fragments or ferrule particles can contaminate the column. Hence, the column ends are pointed towards the floor during cutting.
Legal Information
Shortix is a trademark of Scientific Glass Technology B.V.
Choose from one of the most recent versions:
Certificates of Analysis (COA)