Z765562
Tube rack for Benchmixer™ XL multi-tube vortexer
Holds 50 x 12 mm tubes
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
Recommended Products
feature
Holds 50 x 12 mm tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-12
Related Categories
General description
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables use of 50 x 12 mm tubes in the BenchMixer™ XL heavy-duty hands-free mixing vortex mixer. Ideal for the QuEChERS pesticide analysis method.
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)