Z763721
BeadBug™ prefilled tubes, 2.0 mL capacity
with 0.1 mm Silica glass beads, acid washed
Synonym(s):
homogenization microtubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
colorless tube
conical bottom
screw top cap
silica beads
sterility
non-sterile
feature
graduations: no
packaging
pack of 50 ea
manufacturer/tradename
Benchmark Scientific D1031-01
technique(s)
PCR: suitable
capacity
2 mL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
These prefilled tubes fit the BeadBug™ microtube homogenizer, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads.
The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease.
The stainless steel beads are acid-washed as well.
All tubes are non-skirted with screw-caps
The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease.
The stainless steel beads are acid-washed as well.
All tubes are non-skirted with screw-caps
Legal Information
BeadBug is a trademark of Benchmark Scientific, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service