Z755273
Nunc-Immuno™ standard strip plate module with frame
size 8 wells, polysorb surface, non-sterile
Synonym(s):
module plates, strip plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear polystyrene wells
flat bottom clear polystyrene wells
size:
8 wells
well maximum volume:
400 μm
well surface area:
2.7 cm2
working volume:
350 μL
sterility:
non-sterile
binding type:
(PolySorp® surface)
high binding surface
high binding surface
Recommended Products
material
clear polystyrene
flat bottom clear polystyrene wells
sterility
non-sterile
packaging
case of 60 ea
manufacturer/tradename
Nunc 469078
size
8 wells
well maximum volume
400 μm
well surface area
2.7 cm2
working volume
350 μL
color
none
binding type
(PolySorp® surface)
high binding surface
Related Categories
General description
The Nunc-Immuno strip plates have been developed for ELISA techniques and have an alpha-numeric orientation system. They fit standard equipment and have high optical quality.
Legal Information
Nunc-Immuno is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service