Z722758
Hettich® MIKRO 220/220R centrifuge accessories
for MIKRO 220R Centrifuge, Fixed Angle Rotor, 12 place
All Photos(1)
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
Recommended Products
feature
to hold 12 x 15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1015
parameter
4,146 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MIKRO 220/220R
Related Categories
General description
- compatible with the MIKRO 220R Centrifuge only
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)