Z380296
Jars Nalgene® style 2117
capacity 125 mL
Synonym(s):
laboratory jars, nalgene jars
About This Item
Recommended Products
material
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Joint
Joints threaded neck (70)
description
Autoclavable with closure completely unscrewed
feature
with cap
packaging
pkg of 4
manufacturer/tradename
Nalgene® 2117-0125
bottle diam. × H
64 mm × 74 mm
capacity
125 mL
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)