Key Documents
TMO140360
Nunc® EasyFill Cell Factory System
4 layer, polystyrene, surface area 2528 cm2, sterile, vented, case of 4 ea
Synonym(s):
closed cell system layered storage rack, closed cell system rack
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
material
polystyrene
sterility
sterile
feature
vented
packaging
case of 4 ea
manufacturer/tradename
Nunc 140360
surface area
2528 cm2
binding type
Nunclon Delta surface
General description
The Nunc® EasyFill Cell Factory systems are a space saving, out-of-the-box, multilayer solution and are as easy to use as a T-flask. Scale up your research and commercial-scale applications with confidence. These systems come with a cerrtified Nunclon® Delta cell culture surface to enable consistent performance from lot to lot and format to format. The 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Features and Benefits
  • "Out of the box" system with no accessories required.
  • Large, vented screw closure for easy handling
  • Versatile port design allows for pouring and aseptic filling.
  • Fill and empty five times faster compared to T175 flasks.
  • Enhances productivity by enabling faster operations.
  • Fast start-up with a simple pour of media and cells into the large opening.
  • Versatile port facilitates pouring and aseptic filling techniques.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)