Skip to Content
Merck
All Photos(1)

Key Documents

SAB2105340

Sigma-Aldrich

Anti-JARID2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AI317256, Anti-AU045941, Anti-C79929, Anti-C79931, Anti-Jmj

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

137 kDa

species reactivity

human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JARID2(3720)
mouse ... Jarid2(16468)

Immunogen

Synthetic peptide directed towards the middle region of mouse Jarid2

Biochem/physiol Actions

Jarid2 is a regulator of histone methyltransferase complexes that plays an essential role in embryonic development, including heart and liver development, neural tube fusion process and hematopoiesis.Jarid2 acts by modulating histone methyltransferase activity and promoting the recruitment of histone methyltransferase complexes to their target genes.Jarid2 binds DNA and mediates the recruitment of the PRC2 complex to target genes in embryonic stem cells.Jarid2 does not have histone demethylase activity but regulates activity of various histone methyltransferase complexes. In embryonic stem cells, it associates with the PRC2 complex and inhibits trimethylation of ′Lys-27′ of histone H3 (H3K27me3) by the PRC2 complex, thereby playing a key role in differentiation of embryonic stem cells and normal development. In cardiac cells, it is required to repress expression of cyclin-D1 (CCND1) by activating methylation of ′Lys-9′ of histone H3 (H3K9me) by the GLP1/EHMT1 and G9a/EHMT2 histone methyltransferases.Jarid2 also acts as a transcriptional repressor of ANF via its interaction with GATA4 and NKX2-5.Jarid2 participates in the negative regulation of cell proliferation signaling.

Sequence

Synthetic peptide located within the following region: NASSSCQSTPRKGKTHKHVHNGHVFNGSSRSAREKEPAHKHRSKEATPGK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service