R5026
Four-way microtube rack
blue
Synonym(s):
adapable tube rack, tube rack, tube storage
About This Item
Recommended Products
material
polypropylene (heavy duty)
description
Autoclavable
feature
Holds 12 x 15 mL tubes
Holds 32 x 0.5 mL tubes
Holds 32 x 1.5-2 mL tubes
Holds 4 x 50 mL tubes
manufacturer/tradename
Heathrow Scientific HS29022B
Heathrow Scientific
size
174x95x52 mm
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Racks feature a unique system of tabs and slots, which facilitate easy assembly and sturdy fit.
- A four-way microtube rack can be connected end-to-end or side-to-side to create a large rack to fit various benchtop applications.
- Alphanumeric identification enables quick location and retrieval of tubes.
- The racks can hold 4 × 50 mL conical tubes, 12 × 15 mL conical tubes, 32 × 1.5 mL microtubes, or 32 × 0.5 mL microtubes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service