MAK506
Peroxidase Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Peroxidase Enzyme Activity Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.84
Recommended Products
Quality Level
detection method
colorimetric
fluorometric
relevant disease(s)
cancer; cardiovascular diseases; neurological disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Peroxidase catalyze the following oxidationreduction reactions: ROOR′ + electron donor (2 e-) + 2H+ → ROH + R′OH For many peroxidases the optimal substrate is hydrogen peroxide (H2O2), but others are more active with organic hydroperoxides such as lipid peroxides. In the cell, peroxidases destroy toxic hydroxide radicals that are formed as byproducts during aerobic respiration. The peroxidases represent a large family of enzymes that are found in animals (e.g. myeloperoxidase-like enzymes), plant, fungi, and bacteria (cytochrome-c peroxidase like enzymes such as horseradish peroxidase).
Features and Benefits
- Linear detection range: 2 to 50 U/L for the colorimetric assay and 0.1 to 5 U/L for the fluorometric assay.
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for peroxidase activity determination in biological samples (e.g. plasma, serum, urine, tissue and culture media.)
Principle
The Peroxidase Assay Kit uses H2O2 and an electron donor dye that forms resorufin during the peroxidase reaction. The optical density at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is a direct measure of the enzyme activity.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service