HS120428
TrueNorth® Flatpack PP Freezer Boxes
Holds 100 x vials, natural, pk of 10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.17
Recommended Products
material
polypropylene
feature
Holds 100 x vials
packaging
pk of 10
manufacturer/tradename
Heathrow Scientific HS120428
technique(s)
cryopreservation: suitable
well diam.
17 mm
color
natural
Looking for similar products? Visit Product Comparison Guide
General description
Reusable, economical and moisture resistant plastic freezer boxes.
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service