Skip to Content
Merck
All Photos(4)

Key Documents

HPA015642

Sigma-Aldrich

Anti-AFAP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-110 kDa actin filament-associated protein, Anti-AFAP antibody produced in rabbit, Anti-AFAP-110, Anti-Actin filament-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PEALHYDYIDVEMSASVIQTAKQTFCFMNRRVISANPYLGGTSNGYAHPSGTALHYDDVPCINGSLKGKKPPVASNGVTGKGKTLSSQPKKADPAAVVKRTGSNAAQYKYGKNRVEADAKR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFAP1(60312)

Immunogen

Actin filament-associated protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Actin filament-associated protein 1 is a protein (110 kDa) encoded by the AFAP1 gene in humans. It is found to interact with Src family members and acts as an adaptor protein. It is a tyrosine phosphorylated substrate of Src and is associated with actin filaments integrity (through carboxy terminal interactions) and also can alter actin filament crosslinking protein. It functions as an activator of Src family kinases thereby altering its conformation in response to cellular signals.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72649

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Qian et al.
Oncogene, 16(17), 2185-2195 (1998-06-10)
The SH2 and SH3 binding partner AFAP-110 is a tyrosine phosphorylated substrate of Src. AFAP-110 has been hypothesized to link Src to actin filaments, which may contribute to the effects of Src upon actin filament integrity. However, it has been
Yong Qian et al.
Journal of cellular biochemistry, 91(3), 602-620 (2004-02-03)
AFAP-110 has an intrinsic ability to alter actin filament integrity as an actin filament crosslinking protein. This capability is regulated by a carboxy terminal leucine zipper (Lzip) motif. The Lzip motif facilitates self-association stabilizing the AFAP-110 multimers. Deletion of the
J M Baisden et al.
Oncogene, 20(45), 6607-6616 (2001-10-20)
The actin filament-associated protein of 110 kDa (AFAP-110) is a Src binding partner that represents a potential modulator of actin filament integrity in response to cellular signals. Previous reports have demonstrated that AFAP-110 is capable of directly binding and altering
J M Baisden et al.
Oncogene, 20(44), 6435-6447 (2001-10-19)
The actin filament-associated protein of 110 kDa (AFAP-110) was first identified as an SH3/SH2 binding partner for the nonreceptor tyrosine kinase, Src. Subsequent data have demonstrated that AFAP-110 can interact with other Src family members. AFAP-110 contains additional protein binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service