Skip to Content
Merck
All Photos(5)

Key Documents

HPA006394

Sigma-Aldrich

Anti-PPP1R13B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apoptosis-stimulating of p53 protein 1 antibody produced in rabbit, Anti-Protein phosphatase 1 regulatory subunit 13B antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PNIQKLLYQRFNTLAGGMEGTPFYQPSPSQDFMGTLADVDNGNTNANGNLEELPPAQPTAPLPAEPAPSSDANDNELPSPEPEELICPQTTHQTAEPAEDNNNNVATVPTTEQIPSPVAEAPSPGEEQVPPAPLPPASHPPATSTNK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

PPP1R13B (protein phosphatase 1 regulatory subunit 13B), also called apoptosis-stimulating protein of p53 (ASPP)1, is a member of the ASPP family of proteins, which contains three members namely, ASPP1, ASPP2 and iASPP. Its C-terminal is composed of ankyrin repeats, a SH3 domain and proline-rich sequences. The first 100 residues of its N-terminal contains RAS-association domains (RAD).

Immunogen

Apoptosis-stimulating of p53 protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PPP1R13B (protein phosphatase 1 regulatory subunit 13B) promotes apoptosis in cancer cells, through its interaction with p53, p63 and p73. It is also a transcriptional target of E2F protein. Inactivation of this gene by hypermethylation favors pathogenesis of gestational trophoblastic disease, which is characterized by trophoblastic lesions, such as choriocarcinomas and potentially malignant hydatidiform moles. Its promoter is also aberrantly methylated in acute lymphoblastic leukemia (ALL) resulting in its reduced expression, and this correlates to poor prognosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70509

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Wang et al.
Cell death and differentiation, 20(4), 525-534 (2013-02-09)
RAS mutations occur frequently in human cancer and activated RAS signalling contributes to tumour development and progression. Apart from its oncogenic effects on cell growth, active RAS has tumour-suppressive functions via its ability to induce cellular senescence and apoptosis. RAS
X Agirre et al.
Oncogene, 25(13), 1862-1870 (2005-11-30)
We have analyzed the regulation and expression of ASPP members, genes implicated in the regulation of the apoptotic function of the TP53 tumor-suppressor gene, in acute lymphoblastic leukemia (ALL). Expression of ASPP1 was significantly reduced in ALL and was dependent
Victor C Y Mak et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(4), 522-532 (2010-11-26)
Gestational trophoblastic disease encompasses a spectrum of trophoblastic lesions including true neoplasms such as choriocarcinomas and the potentially malignant hydatidiform moles, which may develop persistent disease requiring chemotherapy. ASPP1, a member of apoptosis-stimulating proteins of p53 (ASPPs), is a proapoptotic
Dian Liu et al.
Cell death & disease, 11(4), 224-224 (2020-04-10)
The apoptosis-stimulating protein of p53 (ASPP) family of proteins can regulate apoptosis by interacting with the p53 family and have been identified to play an important role in cancer progression. Previously, we have demonstrated that ASPP2 downregulation can promote invasion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service