GE27-2066-01
Cytidine 5′-Triphosphate, 100 mM solution (CTP)
Cytiva 27-2066-01
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
NACRES:
NA.52
Recommended Products
form
liquid
packaging
pkg of 25 μmol
manufacturer/tradename
Cytiva 27-2066-01
concentration
100 mM
color
Colorless
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
High purity rNTPs are easy-to-use, time-saving solutions for in vitro transcription assays and/or reactions requiring Ribonucleoside Triphosphates (rNTPs) of high purity. This solution can be used for in vitro transcription or for reactions requiring rNTPs of high purity.
Features and Benefits
- Each lot is tested for ribonuclease contamination to ensure success in your application.
- Supplied in purified water as 100 mM solutions (pH 7.5).
- Extensively purified to yield a minimum of 99% in triphosphate form for solution (98% for lyophilized) to enhance incorporation and yield optimal experimental results.
- Available either as individual products or in a convenient set containing ATP, CTP, GTP, and UTP in separate tubes.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service