Key Documents
EPM1282-9906
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 250 mL Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum
About This Item
UNSPSC Code:
41103816
material
aluminum
feature
dedicated
for 250 mL Erlenmeyer flasks
non-Interchangeable
manufacturer/tradename
Eppendorf® M1282-9906 (Global)
capacity
(40 x 250 mL Erlenmeyer flasks)
General description
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG