EP0030078721
Eppendorf® ep Dualfilter T.I.P.S.® SealMax
PCR clean, sterile, tip volume × L 2-200 μL × 55 mm, yellow, case of 960 tips (10 racks x 96 tips)
Synonym(s):
pipetting tips
About This Item
sterile
Low binding, filter, RNA or DNA free, pcr, low retention etc.
rack of 960 ea (10 racks of 96 tips)
Recommended Products
sterility
sterile (PCR clean/sterile/pyrogen-free)
sterile
feature
PCR clean
Low binding, filter, RNA or DNA free, pcr, low retention etc.
packaging
case of 960 tips (10 racks x 96 tips)
rack of 960 ea (10 racks of 96 tips)
tip volume × L
2-200 μL × 55 mm
color
yellow
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This new Dualfilter closes immediately when it comes into contact with the overpipetted sample without inhibiting valuable material. Additionally drops and splashes are retained by the white layer, the violet layer forms the highly-effective barrier against sample liquid.
Further information and certification on specific sample recovery features, PCR inhibition and aerosol protection are available on your local Eppendorf website.
Product Applications:
- DNA applications (e.g. PCR)
- RNA applications (e.g. Gene expression analysis)
- Protein applications (e.g. Antibody Research)
- Cell Culture applications (e.g. Media)
- Applications with radio actives
- All applications which use aerosol1-binding liquids
Product Features:
- Reliable protection from liquids
- Reliable protection from aerosols1 and biomolecules
- No PCR inhibiting additives
- Protection of your pipette from aggressive liquids when over-aspiration occurs
- Available in PCR clean / Sterile (sterile and pyrogen-free)
- Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service