Skip to Content
Merck
All Photos(1)

Key Documents

CLS3360

Corning® 96 Well Clear Polystyrene Microplate

round bottom clear, Tissue Culture (TC)-treated surface, ampule of 25, sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
round bottom clear
round bottom polystyrene
round wells
size:
96 wells
well volume:
330 μL
well working volume:
75-200 μL
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid: no
skirt
Pricing and availability is not currently available.

material

round bottom clear
round bottom polystyrene
round wells

sterility

sterile

feature

lid: no
skirt

packaging

ampule of 25

manufacturer/tradename

Corning 3360

size

96 wells

well volume

330 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Clear Polystyrene Microplate matrix active group, Round bottom wells with lid has 330 μL total volume.
  • Treated for optimal cell attachment.
  • Non-pyrogenic and sterilized by gamma radiation.
  • Alphanumeric codes for good identification.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service