CLS10035
Solid caps, 33 mm, for Corning® CellBIND® surface hyperflask
disposable screw cap, orange, sterile
Synonym(s):
Corning cell culture flasks, cell culture flasks, cell culture vessels, tissue culture flasks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NA.75
Recommended Products
material
polyethylene
sterility
sterile
feature
closure type screw cap
packaging
case of 4
manufacturer/tradename
Corning 10035
color
orange
Related Categories
General description
This replacement cap can be used on all High Yield PERformance HYPERFlask cell culture vessles (Cat. No. CLS10020, CLS10024, CLS10030, CLS10034).
- 33 mm threaded cap, disposable
- Solid cap is used in closed systems or cell culture applications where venting is not necessary
- Sterilized by gamma irradiation
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service