Skip to Content
Merck
All Photos(1)

Key Documents

IC-01

Reactivity: Anti-Galactocerebroside, monoglactosyldiglyceride psychosine, 93042001

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 93042001-DNA-5UG
pkg of vial of cells 93042001-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3-NS1/1-Ag4.1

Reactivity

Anti-Galactocerebroside, monoglactosyldiglyceride psychosine

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3-NS1/1-Ag4.1 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against galactocerebroside monogalactosyldiglyceride and psychosine. It will react with oligodendrocytes and Schwann cells in cattle, rodents, chick and fish.

Immunogen

Particulate fraction from bovine corpus callosum

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service