B7055
Nalgene® dropper bottle with screw closure
capacity 15 mL, natural bottle, assorted colors cap
Synonym(s):
bottle dropper, dropper bottles
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
low-density polyethylene
natural bottle
round bottle
Joint
Joints threaded neck (15)
packaging
pack of 25 ea
manufacturer/tradename
Nalgene® 2752-9050
capacity
15 mL
color
assorted colors cap
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
- The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
- Chemically resistant to most reagents.
- LDPE clear transparent bottles for easy sample visualization.
- Leak-proof closure - prevents sample loss.
- Control dispensing tip ensures a secure fit.
- Suitable for use in biotech, diagnostic and pharmaceutical applications.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service