AXYT350CRS
Pipette tips
size 300 μL, pkg of 5x960tips/cs, racked, clear, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
size:
300 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc T-350-C-R-S
Corning
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-350-C-R-S
Corning
packaging
pkg of 5x960tips/cs
size
300 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Made up of polypropylene
- Fine-point pipette tip
- Nonpyrogenic in nature and RNase-/DNase-free
- Compatible with a wide range of single and multi-channel pipettors
- Requires less force to load and eject with a secure fit due to flexible walls and a series of internal sealing rings
Other Notes
Browse all pipette tips
Legal Information
Corning is a registered trademark of Corning, Inc.
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service